missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Papanicolaou Stain, EA-65 Formula, Ricca Chemical
Counterstain for use with Hematoxylin Stains in Papanicolaou Staining
Supplier: Ricca Chemical Company 550532
Chemical Identifiers
| 91.0, 6.07, 0.27, 0.24, 0.06, 0.02 | |
| 95.0, 6.32, 0.28, 0.25, 0.06, 0.02 | |
| 7732-18-5, 17372-87-1, 12067-99-1, 10114-58-6, 5141-20-8 | |
| Papanicolaou Stain, EA-65 Formula |
Specifications
| Papanicolaou Stain | |
| 91.0, 6.07, 0.27, 0.24, 0.06, 0.02 | |
| 7732-18-5 , 17372-87-1 , 12067-99-1 , 10114-58-6 , 5141-20-8 | |
| Green |
| EA-65 Formula | |
| 95.0, 6.32, 0.28, 0.25, 0.06, 0.02 | |
| Natural Poly Bottle | |
| 1 L |