missing translation for 'onlineSavingsMsg'
Learn More

Papain, Powder, Purified, Spectrum™ Chemical

Catalog No. 18604979
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604979 500 g Poly Bottle
18604978 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18604979 Supplier Spectrum Chemical Mfg Cor Supplier No. PA125500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PA125, 9001-73-4

Spectrum™ Chemical Papain, Powder, Purified , is a cysteine protease enzyme the is present in the papaya fruit. It is traditionnally used to break down meat fibers. It has more recently been used to dissociate cells in the early steps of cell culture preparation.

Chemical Identifiers

CAS 9001-73-4

Specifications

Grade Purified
Packaging Poly Bottle
Quantity 500 g