missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Papain, Carica Papaya Latex

Catalog No. p-7082291
Change view
Click to view available options
Quantity:
25 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61875MB 25 mg
AAJ61875MC 100 mg
AAJ6187503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ61875MB Supplier Thermo Scientific Chemicals Supplier No. J61875MB
Only null left

A cysteine protease; breaks down intercellular matrix of cartilage | CAS: 9001-73-423 kd g/mol

Papain, from papaya latex, has been used to digest carotid artery smooth muscle cells. Papain is used in dissecting solutions. It is used to produce Fab fragments of antibodies. It is used for cell dissociation since it has been shown to be more effective and less damaging with certain tissues.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9001-73-4
Synonym EC 3.4.22.2

Specifications

For Use With (Application) A cysteine protease; breaks down intercellular matrix of cartilage
Molecular Weight (g/mol) 23 kd
Content And Storage 2°C to 8°C
Form Lyophilized powder,23 kDa
Quantity 25 mg
Product Type Protease
EINECSNumber 232-627-2
Recommended Storage Keep cold

RUO – Research Use Only