missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pantoprazole Sulfide 98.0+%, TCI America™
$51.98 - $155.93
Chemical Identifiers
| CAS | 102625-64-9 |
|---|---|
| Molecular Formula | C16H15F2N3O3S |
| Molecular Weight (g/mol) | 367.37 |
| MDL Number | MFCD07368273 |
| InChI Key | UKILEIRWOYBGEJ-UHFFFAOYSA-N |
| Synonym | pantoprazole sulfide, 5-difluoromethoxy-2-3,4-dimethoxy-2-pyridinyl methyl thio-1h-benzimidazole, 5-difluoromethoxy-2-3,4-dimethoxypyridin-2-yl methyl thio-1h-benzo d imidazole, unii-bwz6x03hib, bwz6x03hib, 5-difluoromethoxy-2-3,4-dimethoxypyridin-2-yl methyl sulfanyl-1h-1,3-benzodiazole, 5-difluoromethoxy-2-3,4-dimethoxypyridin-2-yl methyl thio-1h-benzimidazole, pantoprazole thioether, pantoprazole related compound b |
| PubChem CID | 9799341 |
| IUPAC Name | 6-(difluoromethoxy)-2-{[(3,4-dimethoxypyridin-2-yl)methyl]sulfanyl}-1H-1,3-benzodiazole |
| SMILES | COC1=C(OC)C(CSC2=NC3=CC=C(OC(F)F)C=C3N2)=NC=C1 |
Chemical Identifiers
| 102625-64-9 | |
| 367.37 | |
| UKILEIRWOYBGEJ-UHFFFAOYSA-N | |
| 9799341 | |
| COC1=C(OC)C(CSC2=NC3=CC=C(OC(F)F)C=C3N2)=NC=C1 |
| C16H15F2N3O3S | |
| MFCD07368273 | |
| pantoprazole sulfide, 5-difluoromethoxy-2-3,4-dimethoxy-2-pyridinyl methyl thio-1h-benzimidazole, 5-difluoromethoxy-2-3,4-dimethoxypyridin-2-yl methyl thio-1h-benzo d imidazole, unii-bwz6x03hib, bwz6x03hib, 5-difluoromethoxy-2-3,4-dimethoxypyridin-2-yl methyl sulfanyl-1h-1,3-benzodiazole, 5-difluoromethoxy-2-3,4-dimethoxypyridin-2-yl methyl thio-1h-benzimidazole, pantoprazole thioether, pantoprazole related compound b | |
| 6-(difluoromethoxy)-2-{[(3,4-dimethoxypyridin-2-yl)methyl]sulfanyl}-1H-1,3-benzodiazole |
Specifications
| 102625-64-9 | |
| C16H15F2N3O3S | |
| 1 g | |
| UKILEIRWOYBGEJ-UHFFFAOYSA-N | |
| 6-(difluoromethoxy)-2-{[(3,4-dimethoxypyridin-2-yl)methyl]sulfanyl}-1H-1,3-benzodiazole | |
| 9799341 | |
| ≥98.0% (HPLC,T) | |
| Pantoprazole Sulfide |
| 118°C | |
| MFCD07368273 | |
| pantoprazole sulfide, 5-difluoromethoxy-2-3,4-dimethoxy-2-pyridinyl methyl thio-1h-benzimidazole, 5-difluoromethoxy-2-3,4-dimethoxypyridin-2-yl methyl thio-1h-benzo d imidazole, unii-bwz6x03hib, bwz6x03hib, 5-difluoromethoxy-2-3,4-dimethoxypyridin-2-yl methyl sulfanyl-1h-1,3-benzodiazole, 5-difluoromethoxy-2-3,4-dimethoxypyridin-2-yl methyl thio-1h-benzimidazole, pantoprazole thioether, pantoprazole related compound b | |
| COC1=C(OC)C(CSC2=NC3=CC=C(OC(F)F)C=C3N2)=NC=C1 | |
| 367.37 | |
| 367.37 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator