accessibility menu, dialog, popup

UserName

CAS RN 102625-64-9

H3CONSNHNOFFOCH3

CAS RN 102625-64-9

IUPAC Name: 6-(difluoromethoxy)-2-{[(3,4-dimethoxypyridin-2-yl)methyl]sulfanyl}-1H-1,3-benzodiazole
Synonyms: 5-(difluoromethoxy)-2-{[(3,4-dimethoxypyridin-2-yl)methyl]sulfanyl}-3H-1,3-benzodiazole
Molecular Weight (g/mol): 367.37
Molecular Formula: C16H15F2N3O3S
InChi Key: UKILEIRWOYBGEJ-UHFFFAOYSA-N
SMILES: COC1=C(OC)C(CSC2=NC3=CC=C(OC(F)F)C=C3N2)=NC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results