missing translation for 'onlineSavingsMsg'
Learn More

Pantoprazole Sulfide 98.0+%, TCI America™

Catalog No. P20661G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20661G 1 g
P20665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P20661G Supplier TCI America Supplier No. P20661G
Only null left

Chemical Identifiers

CAS 102625-64-9
Molecular Formula C16H15F2N3O3S
Molecular Weight (g/mol) 367.37
MDL Number MFCD07368273
InChI Key UKILEIRWOYBGEJ-UHFFFAOYSA-N
Synonym pantoprazole sulfide, 5-difluoromethoxy-2-3,4-dimethoxy-2-pyridinyl methyl thio-1h-benzimidazole, 5-difluoromethoxy-2-3,4-dimethoxypyridin-2-yl methyl thio-1h-benzo d imidazole, unii-bwz6x03hib, bwz6x03hib, 5-difluoromethoxy-2-3,4-dimethoxypyridin-2-yl methyl sulfanyl-1h-1,3-benzodiazole, 5-difluoromethoxy-2-3,4-dimethoxypyridin-2-yl methyl thio-1h-benzimidazole, pantoprazole thioether, pantoprazole related compound b
PubChem CID 9799341
IUPAC Name 6-(difluoromethoxy)-2-{[(3,4-dimethoxypyridin-2-yl)methyl]sulfanyl}-1H-1,3-benzodiazole
SMILES COC1=C(OC)C(CSC2=NC3=CC=C(OC(F)F)C=C3N2)=NC=C1

Specifications

Melting Point 118°C
Quantity 1 g
Formula Weight 367.37
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pantoprazole Sulfide
TSCA No
Recommended Storage Refrigerator