missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ PANCREATIN 4X from Porcine Pancreas

Used to treat conditions in which pancreatic secretions are deficient, such as surgical pancreatectomy, pancreatitis and cystic fibrosis.

$227.69

Specifications

Color Tan
For Use With (Application) Convert starch into soluble carbohydrates
Content And Storage 0°C
Product Type Proteins, Enzymes & Peptides
Form Powder
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP219397580
View Documents Promotion Details
MP Biomedicals™
0219397580
100 g
Each for $227.69
Only null left
 
Description

Description

Applications: Pancreatin is used to treat conditions in which pancreatic secretions are deficient, such as surgical pancreatectomy, pancreatitis and cystic fibrosis. It has been claimed to help with food allergies, celiac disease, autoimmune disease, cancer and weight loss.
Product Description: Pancreatin is a mixture of digestive enzymes produced by the exocrine cells of the pancreas. It is composed of amylase, lipase and protease. The trypsin found in pancreatin works to hydrolyze proteins into oligopeptides; amylase hydrolyzes starches into oligosaccharides and the disaccharide maltose; and lipase hydrolyzes triglycerides into fatty acids and glycerols.
Biochem/physiol Actions: Pancreatin will convert not less than 25 times its weight of potato starch into soluble carbohydrates in 5 minutes in water at 40 °C, will digest not less than 25 times its weight of casein in 60 minutes at pH 7.5 at 40 °C and will release not less than 2 microequivalents of acid per min per mg pancreatin from olive oil at pH 9.0 at 37 °C.
Specifications

Specifications

Tan
0°C
Powder
Convert starch into soluble carbohydrates
Proteins, Enzymes & Peptides
Videos
SDS
Documents

Documents