Learn More
Palmitoyl chloride, 98%
CAS: 112-67-4 | C16H31ClO | 274.873 g/mol
Supplier: Thermo Scientific Chemicals A1381222
Description
Palmitoyl chloride is a compound which has been used as a polymeric carrier for plasmid DNA delivery to 293T cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPalmitoyl chloride is a compound which has been used as a polymeric carrier for plasmid DNA delivery to 293T cells.
Solubility
Soluble in ether.Decomposes in water and alcohol.
Notes
Moisture Sensitive. Desiccate at room temperature. Store under inert gas. Incompatible with Strong oxidizing agents, Bases, Alcohols.
Specifications
| Palmitoyl chloride | |
| ∼9°C to 12°C | |
| 174°C (0.5 mmHg) | |
| C16H31ClO | |
| CH3(CH2)14COCl | |
| 100 g | |
| 972409 | |
| palmitoyl chloride, palmitic acid chloride, palmitoylchloride, unii-7642r2josb, hexadecanoyl-chloride, palm acetyl chlorine 14 acetyl chlorine, hexadecanoic acid, chloride, palmitic chloride, hexadecanoylchloride, n-hexadecanoyl chloride | |
| ARBOVOVUTSQWSS-UHFFFAOYSA-N | |
| hexadecanoyl chloride | |
| 8206 | |
| 98% |
| 112-67-4 | |
| 0.906 | |
| 182°C (359°F) | |
| 1.452 | |
| MFCD00000742 | |
| UN3265 | |
| Moisture sensitive | |
| Soluble in ether.Decomposes in water and alcohol. | |
| CCCCCCCCCCCCCCCC(=O)Cl | |
| 274.873 | |
| 274.88 |
Chemical Identifiers
| 112-67-4 | |
| 274.873 | |
| ARBOVOVUTSQWSS-UHFFFAOYSA-N | |
| 8206 | |
| CCCCCCCCCCCCCCCC(=O)Cl |
| C16H31ClO | |
| MFCD00000742 | |
| palmitoyl chloride, palmitic acid chloride, palmitoylchloride, unii-7642r2josb, hexadecanoyl-chloride, palm acetyl chlorine 14 acetyl chlorine, hexadecanoic acid, chloride, palmitic chloride, hexadecanoylchloride, n-hexadecanoyl chloride | |
| hexadecanoyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
H314-H317
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 203-996-7
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only