missing translation for 'onlineSavingsMsg'
Learn More

Palmitoyl chloride, 98%

Catalog No. p-7021467
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1381222 100 g
AAA1381236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1381222 Supplier Thermo Scientific Chemicals Supplier No. A1381222

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 112-67-4 | C16H31ClO | 274.873 g/mol

Palmitoyl chloride is a compound which has been used as a polymeric carrier for plasmid DNA delivery to 293T cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Palmitoyl chloride is a compound which has been used as a polymeric carrier for plasmid DNA delivery to 293T cells.

Solubility
Soluble in ether.Decomposes in water and alcohol.

Notes
Moisture Sensitive. Desiccate at room temperature. Store under inert gas. Incompatible with Strong oxidizing agents, Bases, Alcohols.

Chemical Identifiers

CAS 112-67-4
Molecular Formula C16H31ClO
Molecular Weight (g/mol) 274.873
MDL Number MFCD00000742
InChI Key ARBOVOVUTSQWSS-UHFFFAOYSA-N
Synonym palmitoyl chloride, palmitic acid chloride, palmitoylchloride, unii-7642r2josb, hexadecanoyl-chloride, palm acetyl chlorine 14 acetyl chlorine, hexadecanoic acid, chloride, palmitic chloride, hexadecanoylchloride, n-hexadecanoyl chloride
PubChem CID 8206
IUPAC Name hexadecanoyl chloride
SMILES CCCCCCCCCCCCCCCC(=O)Cl

Specifications

Melting Point ∼9°C to 12°C
Density 0.906
Boiling Point 174°C (0.5 mmHg)
Flash Point 182°C (359°F)
Refractive Index 1.452
Linear Formula CH3(CH2)14COCl
Quantity 100 g
UN Number UN3265
Beilstein 972409
Sensitivity Moisture sensitive
Solubility Information Soluble in ether.Decomposes in water and alcohol.
Formula Weight 274.88
Percent Purity 98%
Chemical Name or Material Palmitoyl chloride
Show More Show Less
Hazard Category H314-H317
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 203-996-7
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only