missing translation for 'onlineSavingsMsg'
Learn More

Palmitic Acid, Approx. 92%, Spectrum™ Chemical

Chemical Identifiers

CAS 57-10-3
Molecular Weight (g/mol) 256.42 g/mol
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18612119
View Documents Promotion Details
N/A 500 g Poly Pail N/A
Only null left
 
18612117
View Documents Promotion Details
N/A 2.5 kg Poly Pail N/A
Only null left
 
18612116
View Documents Promotion Details
N/A 12 kg Poly Pail N/A
Only null left
 
18612118
View Documents Promotion Details
N/A 45 kg Poly Pail N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

57-10-3
256.42 g/mol

Specifications

57-10-3
59° to 64°C
256.42 g/mol
Reagent
Palmitic Acid
1
350°C
∼92%
Crystals or Flakes
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.