missing translation for 'onlineSavingsMsg'
Learn More

Palmitic Acid, Approx. 92%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor PA1102.5KG

Quantity 2.5 kg
Packaging Poly Pail

 View more versions of this product

Catalog No. 18612117


Only null left

Description

Description

Specifications

Chemical Identifiers

57-10-3
256.42 g/mol

Specifications

Palmitic Acid
1
350°C
2.5 kg
∼92%
Crystals or Flakes
57-10-3
59° to 64°C
Poly Pail
256.42 g/mol
Reagent
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions