missing translation for 'onlineSavingsMsg'
Learn More

Palladium on activated carbon, 10% Pd, (50% wet with water for safety), unreduced

Catalog No. AC422980250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC422980250 25 g Glass bottle
AC422981000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC422980250 Supplier Thermo Scientific Chemicals Supplier No. 422980250

CAS: 7440-05-3 | Pd | 106.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-05-3 , 7732-18-5 , 7440-44-0
Molecular Formula Pd
Molecular Weight (g/mol) 106.42
MDL Number MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
Synonym black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
PubChem CID 23938
ChEBI CHEBI:33363
IUPAC Name palladium
SMILES [Pd]

Specifications

Color Black
Quantity 25 g
Packaging Glass bottle
Form Powder
Linear Formula Pd
Fieser 01,778; 04,365; 05,498; 06,443; 07,274; 15,13; 16,356
Formula Weight 106.42
Percent Purity 10% (Pd)
Chemical Name or Material Palladium on activated carbon

RUO – Research Use Only