missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Palatinose hydrate, 98+%

CAS: 343336-76-5 | C12H22O11 | 342.30 g/mol

Supplier:  Thermo Scientific Chemicals J6009114

 View more versions of this product

Catalog No. AAJ6009114


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Palatinose hydrate
122°C to 124°C
C12H22O11
25 g
14,5182
PVXPPJIGRGXGCY-VVLSOAGWNA-N
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[[(2R,3S,4S)-3,4,5-trihydroxy-5-(hydroxymethyl)oxolan-2-yl]methoxy]oxane-3,4,5-triol;hydrate
52994241
≥98%
343336-76-5
White
MFCD00076094
1757215
6-o-alpha-d-glucopyranosyl-beta-d-fructofuranose, beta-palatinose, beta-f-palatinose, alpha-d-glucopyranose-1->6-beta-d-fructofuranose, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-2r,3s,4s,5r-3,4,5-trihydroxy-5-hydroxymethyl oxolan-2-yl methoxy oxane-3,4,5-triol
OC[C@H]1O[C@H](OC[C@H]2O[C@@](O)(CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
342.30
342.3
Powder

Chemical Identifiers

343336-76-5
342.30
PVXPPJIGRGXGCY-VVLSOAGWNA-N
52994241
OC[C@H]1O[C@H](OC[C@H]2O[C@@](O)(CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
C12H22O11
MFCD00076094
6-o-alpha-d-glucopyranosyl-beta-d-fructofuranose, beta-palatinose, beta-f-palatinose, alpha-d-glucopyranose-1->6-beta-d-fructofuranose, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-2r,3s,4s,5r-3,4,5-trihydroxy-5-hydroxymethyl oxolan-2-yl methoxy oxane-3,4,5-triol
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[[(2R,3S,4S)-3,4,5-trihydroxy-5-(hydroxymethyl)oxolan-2-yl]methoxy]oxane-3,4,5-triol;hydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 237-282-1

RTECSNumber : LS7175420

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only