accessibility menu, dialog, popup

UserName

CAS RN 343336-76-5

IUPAC Name: (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4S,5S)-3,4,5-trihydroxy-5-(hydroxymethyl)oxolan-2-yl]methoxy}oxane-3,4,5-triol
Synonyms: (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4S,5S)-3,4,5-trihydroxy-5-(hydroxymethyl)oxolan-2-yl]methoxy}oxane-3,4,5-triol
Molecular Weight (g/mol): 342.3
Molecular Formula: C12H22O11
InChi Key: PVXPPJIGRGXGCY-VVLSOAGWNA-N
SMILES: OC[C@H]1O[C@H](OC[C@H]2O[C@@](O)(CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (2)
  • (3)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)