missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Packing Reservoirs for Non-Jacketed LC columns

Catalog No. 111007729
Change view
Click to view available options
Size (Metric):
2,000 mL
Description:
Column I.D.: 4.8 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (Metric) Description
111007729 2,000 mL Column I.D.: 4.8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111007729 Supplier MilliporeSigma™ Supelco™ Supplier No. R62581EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Supplied complete with all fittings necessary for connection to threaded columns. The top has 1/4 -28 fitting.

Specifications

Size (Metric) 2,000 mL
For Use With (Application) Liquid Chromatography
Description Column I.D.: 4.8 cm