missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Pack of 12 Sterile Wide Mouth HDPE Reagent Bottles with Polypropylene Caps
Pack of 12 sterile wide-mouth HDPE reagent bottles with polypropylene caps are leakproof, chemically resistant, and easy to fill and pour, making them ideal for secure liquid storage and handling in lab or industrial environments.
$64.57 - $75.39
Specifications
| Color | Translucent |
|---|---|
| Diameter (Metric) Inner Neck | 21.6 mm |
| Material | HDPE |
| No. per Pack | 12 Pk. |
| Sterility | Sterile |
Description
- Sterile HDPE bottles: Pack of 12 reagent bottles made from HDPE with polypropylene caps. Sterile and ideal for secure reagent storage and lab applications.
- Wide mouth design: Features a wide-mouth opening for easy filling, pouring, and handling in laboratory environments.
- Chemical resistance: Designed to withstand exposure to a wide range of chemicals, making these bottles suitable for diverse industrial and lab uses. These bottles offer excellent chemical resistance and a -100°C to +120°C temperature range.
- Leakproof tested: Tested for leakage prevention under high pressure to ensure safe storage and transport of liquids and reagents.
- Translucent and durable: Manufactured in a controlled environment for reliability, these translucent bottles provide visibility of contents and ease of use.
Specifications
| Translucent | |
| HDPE | |
| Sterile | |
| No | |
| Reagent Bottle |
| 21.6 mm | |
| 12 Pk. | |
| Yes | |
| Wide | |
| Not Autoclavable |