missing translation for 'onlineSavingsMsg'
Learn More

Eisco Labs Pack of 12 Sterile Wide Mouth HDPE Reagent Bottles with Polypropylene Caps

Catalog No. S000122
Change view
Click to view available options
Capacity (Metric):
30 mL
60 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S000122 30 mL
S000123 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S000122 Supplier Eisco Labs Supplier No. CH5471PK12

Pack of 12 sterile wide-mouth HDPE reagent bottles with polypropylene caps are leakproof, chemically resistant, and easy to fill and pour, making them ideal for secure liquid storage and handling in lab or industrial environments.

  • Sterile HDPE bottles: Pack of 12 reagent bottles made from HDPE with polypropylene caps. Sterile and ideal for secure reagent storage and lab applications.
  • Wide mouth design: Features a wide-mouth opening for easy filling, pouring, and handling in laboratory environments.
  • Chemical resistance: Designed to withstand exposure to a wide range of chemicals, making these bottles suitable for diverse industrial and lab uses. These bottles offer excellent chemical resistance and a -100°C to +120°C temperature range.
  • Leakproof tested: Tested for leakage prevention under high pressure to ensure safe storage and transport of liquids and reagents.
  • Translucent and durable: Manufactured in a controlled environment for reliability, these translucent bottles provide visibility of contents and ease of use.

Specifications

Capacity (Metric) 30 mL
Capacity (English) 1 oz.
Color Translucent
Diameter (Metric) Inner Neck 21.6 mm
Material HDPE
No. per Pack 12 Pk.
Sterility Sterile
Closure Included Yes
Graduated No
Mouth Wide
Height (Metric) 62 mm
Product Type Reagent Bottle
Diameter (Metric) 34.8 mm
Autoclavable Not Autoclavable
Show More Show Less