missing translation for 'onlineSavingsMsg'
Learn More

Ferric Ammonium Sulfate, Tenth-Normal (0.1 N) Volumetric Solution, USP, Spectrum™ Chemical

Catalog No. 18602665
Change view
Click to view available options
Quantity:
1 L
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602665 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18602665 Supplier Spectrum Chemical Mfg Cor Supplier No. F1611LT
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

F-161, 7783-83-7

Spectrum™ Chemical Ferric Ammonium Sulfate, Tenth-Normal (0.1 N), USP Volumetric Solution - Spectrum USP Volumetric Solutions are also known as Standard Solutions, are solutions of reagents of known concentration intended primarily for use in quantitative determinations.

Chemical Identifiers

CAS 7783-83-7 , 7732-18-5
Molecular Formula FeH28NO20S2
Molecular Weight (g/mol) 482.18
InChI Key LCPUDZUWZDSKMX-UHFFFAOYSA-K
IUPAC Name iron(3+) ammonium dodecahydrate disulfate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

CAS Max % 4.8%, 94.6%
Physical Form Liquid
Quantity 1 L
Grade Ungraded
Concentration or Composition (by Analyte or Components) 0.1 N +/-0.002
Packaging Amber Glass Bottle
Chemical Name or Material Ferric Ammonium Sulfate USP Volumetric Solution