missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals p21-Activated Kinase Inhibitor III, IPA-3

Acts as an allosteric inhibitor of Pak1

$230.86 - $388.36

Chemical Identifiers

CAS 42521-82-4
Molecular Formula C20H14O2S2
Synonym PAK Inhibitor III; 1,1′-Disulfanediylbis(naphthalen-2-ol)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ65848MA
View Documents Promotion Details
Thermo Scientific Chemicals
J65848MA
10 mg
Each for $388.36
Only null left
 
AAJ65848LB0
View Documents Promotion Details
Thermo Scientific Chemicals
J65848LB0
5 mg
Each for $230.86
Only null left
 
Description

Description

Acts as an allosteric inhibitor of Pak1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

42521-82-4
C20H14O2S2
PAK Inhibitor III; 1,1′-Disulfanediylbis(naphthalen-2-ol)

Specifications

Yellow
Acts as an allosteric inhibitor of Pak1
C20H14O2S2
PAK Inhibitor III; 1,1′-Disulfanediylbis(naphthalen-2-ol)
Soluble in DMSO or ethanol; Insoluble in water
Solid
OC1=CC=C2C=CC=CC2=C1SSC1=C(O)C=CC2=CC=CC=C12
350.45
-30°C to -10°C
42521-82-4
350.50g/mol
Light Sensitive
Signal Transduction Reagent
RFAXLXKIAKIUDT-UHFFFAOYSA-N
1-[(2-hydroxynaphthalen-1-yl)disulfanyl]naphthalen-2-ol
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

RUO – Research Use Only