missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals p21-Activated Kinase Inhibitor III, IPA-3
Acts as an allosteric inhibitor of Pak1
Supplier: Thermo Scientific Chemicals J65848LB0
| Quantity | 5 mg |
|---|
Description
Acts as an allosteric inhibitor of Pak1
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 42521-82-4 | |
| C20H14O2S2, C20H14O2S2 | |
| PAK Inhibitor III; 1,1′-Disulfanediylbis(naphthalen-2-ol) |
Specifications
| 5 mg | |
| -30°C to -10°C | |
| 42521-82-4 | |
| 350.50g/mol | |
| Light Sensitive | |
| Signal Transduction Reagent | |
| RFAXLXKIAKIUDT-UHFFFAOYSA-N | |
| 1-[(2-hydroxynaphthalen-1-yl)disulfanyl]naphthalen-2-ol |
| Yellow | |
| Acts as an allosteric inhibitor of Pak1 | |
| C20H14O2S2, C20H14O2S2 | |
| PAK Inhibitor III; 1,1′-Disulfanediylbis(naphthalen-2-ol) | |
| Soluble in DMSO or ethanol; Insoluble in water | |
| Solid | |
| OC1=CC=C2C=CC=CC2=C1SSC1=C(O)C=CC2=CC=CC=C12 | |
| 350.45 |
Safety and Handling
Recommended Storage : Store at -20°C