missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Xylylene Dithiocyanate 98.0+%, TCI America™
$233.00 - $719.52
Chemical Identifiers
| CAS | 1014-99-9 |
|---|---|
| Molecular Formula | C10H8N2S2 |
| Molecular Weight (g/mol) | 220.308 |
| InChI Key | ZMFLUYPLYCZQDR-UHFFFAOYSA-N |
| PubChem CID | 4460705 |
| IUPAC Name | [4-(thiocyanatomethyl)phenyl]methyl thiocyanate |
| SMILES | C1=CC(=CC=C1CSC#N)CSC#N |
Chemical Identifiers
| 1014-99-9 | |
| 220.308 | |
| 4460705 | |
| C1=CC(=CC=C1CSC#N)CSC#N |
| C10H8N2S2 | |
| ZMFLUYPLYCZQDR-UHFFFAOYSA-N | |
| [4-(thiocyanatomethyl)phenyl]methyl thiocyanate |
Specifications
| 1014-99-9 | |
| White | |
| 5 g | |
| alpha,alpha′C-Dithiocyanato-p-xylene | |
| C1=CC(=CC=C1CSC#N)CSC#N | |
| 220.308 | |
| 220.31 | |
| Crystalline Powder |
| 137°C | |
| C10H8N2S2 | |
| 2811 | |
| ZMFLUYPLYCZQDR-UHFFFAOYSA-N | |
| [4-(thiocyanatomethyl)phenyl]methyl thiocyanate | |
| 4460705 | |
| ≥98.0% (GC) | |
| p-Xylylene Dithiocyanate |
Safety and Handling
TSCA : No