accessibility menu, dialog, popup

UserName

CAS RN 1014-99-9

IUPAC Name: [({4-[(cyanosulfanyl)methyl]phenyl}methyl)sulfanyl]carbonitrile
Synonyms: ({4-[(cyanosulfanyl)methyl]phenyl}methyl)sulfanylcarbonitrile
Molecular Weight (g/mol): 220.31
Molecular Formula: C10H8N2S2
InChi Key: ZMFLUYPLYCZQDR-UHFFFAOYSA-N
SMILES: N#CSCC1=CC=C(CSC#N)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results