missing translation for 'onlineSavingsMsg'
Learn More

p-Xylylene Dithiocyanate 98.0+%, TCI America™

Catalog No. X00605G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X00605G 5 g
X006025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. X00605G Supplier TCI America Supplier No. X00605G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1014-99-9
Molecular Formula C10H8N2S2
Molecular Weight (g/mol) 220.308
InChI Key ZMFLUYPLYCZQDR-UHFFFAOYSA-N
PubChem CID 4460705
IUPAC Name [4-(thiocyanatomethyl)phenyl]methyl thiocyanate
SMILES C1=CC(=CC=C1CSC#N)CSC#N

Specifications

Melting Point 137°C
Color White
Quantity 5 g
UN Number 2811
Synonym alpha,alpha′C-Dithiocyanato-p-xylene
Formula Weight 220.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material p-Xylylene Dithiocyanate