missing translation for 'onlineSavingsMsg'
Learn More

p-Xylylene dichloride, 98%

Catalog No. p-7021579
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1397622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1397622 Supplier Thermo Scientific Chemicals Supplier No. A1397622

CAS: 623-25-6 | C8H8Cl2 | 175.052 g/mol

p-Xylylene dichloride is a monomer used in preparation of poly(p-phenylene vinylene(PPV) nanotubes and nanorods by chemical vapor deposition (CVD) polymerization method. It was used in a study to investigate the effect of CVD monomer selection and reaction conditions on the resulting PPV polymer layer composition and luminescence properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Xylylene dichloride is a monomer used in preparation of poly(p-phenylene vinylene(PPV) nanotubes and nanorods by chemical vapor deposition (CVD) polymerization method. It was used in a study to investigate the effect of CVD monomer selection and reaction conditions on the resulting PPV polymer layer composition and luminescence properties.

Solubility
Insoluble in water

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, bases and active metals.

Chemical Identifiers

CAS 623-25-6
Molecular Formula C8H8Cl2
Molecular Weight (g/mol) 175.052
MDL Number MFCD00000920
InChI Key ZZHIDJWUJRKHGX-UHFFFAOYSA-N
Synonym 1,4-bis chloromethyl benzene, alpha,alpha'-dichloro-p-xylene, p-xylylene dichloride, p-xylylene chloride, benzene, 1,4-bis chloromethyl, p-bis chloromethyl benzene, benzene, p-bis chloromethyl, a,a'-dichloro-p-xylene, xylylene dichloride, ccris 1772
PubChem CID 12171
IUPAC Name 1,4-bis(chloromethyl)benzene
SMILES C1=CC(=CC=C1CCl)CCl

Specifications

Melting Point 98°C to 102°C
Density 1.417
Boiling Point 254°C
Quantity 100 g
UN Number UN2928
Beilstein 774706
Solubility Information Insoluble in water
Formula Weight 175.06
Percent Purity 98%
Chemical Name or Material p-Xylylene dichloride
Hazard Category H302-H314-H330
Hazard Statement GHS H Statement
H330-H314-H318-H302
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.
EINECSNumber 210-782-7
RTECSNumber ZE4100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only