accessibility menu, dialog, popup

UserName

ClCl

CAS RN 623-25-6

IUPAC Name: 1,4-bis(chloromethyl)benzene
Synonyms: xylylene dichloride
Molecular Weight (g/mol): 175.05
Molecular Formula: C8H8Cl2
InChi Key: ZZHIDJWUJRKHGX-UHFFFAOYSA-N
SMILES: ClCC1=CC=C(CCl)C=C1
Boiling Point 
  • (3)
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (3)
  • (2)
  • (2)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (3)
  • (1)
  • (3)
  • (5)
  • (2)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)