missing translation for 'onlineSavingsMsg'
Learn More

p-Xylylene dibromide, 97%

CAS: 623-24-5 | C8H8Br2 | 263.96 g/mol

Supplier:  Thermo Scientific Chemicals A1393918

 View more versions of this product

Catalog No. AAA1393918


Only null left
Explore available promotions

Description

Description

It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Soluble in water (react), and toluene.

Notes
Incompatible with alcohols, bases, amines, oxidizing agents. Avoid moisture.
Specifications

Specifications

p-Xylylene dibromide
144°C to 148°C
245°C
MFCD00000182
UN3261
1,4-bis bromomethyl benzene, p-xylylene dibromide, alpha,alpha'-dibromo-p-xylene, benzene, 1,4-bis bromomethyl, p-xylylene bromide, p-bromomethyl benzene, 1,4-dibromomethylbenzene, p-bis bromomethyl benzene, 1,4-bis-bromomethyl-benzene, ccris 1775
RBZMSGOBSOCYHR-UHFFFAOYSA-N
1,4-bis(bromomethyl)benzene
69335
97%
623-24-5
2.012
C8H8Br2
50 g
1100019
Soluble in water (react),and toluene.
C1=CC(=CC=C1CBr)CBr
263.96
263.96

Chemical Identifiers

623-24-5
263.96
RBZMSGOBSOCYHR-UHFFFAOYSA-N
69335
C1=CC(=CC=C1CBr)CBr
C8H8Br2
MFCD00000182
1,4-bis bromomethyl benzene, p-xylylene dibromide, alpha,alpha'-dibromo-p-xylene, benzene, 1,4-bis bromomethyl, p-xylylene bromide, p-bromomethyl benzene, 1,4-dibromomethylbenzene, p-bis bromomethyl benzene, 1,4-bis-bromomethyl-benzene, ccris 1775
1,4-bis(bromomethyl)benzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P363-P501c

H314-H318

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 210-781-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only