missing translation for 'onlineSavingsMsg'
Learn More

p-Xylylene dibromide, 97%

Catalog No. p-7021553
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1393909 10 g
AAA1393918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1393909 Supplier Thermo Scientific Chemicals Supplier No. A1393909

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 623-24-5 | C8H8Br2 | 263.96 g/mol

It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Soluble in water (react), and toluene.

Notes
Incompatible with alcohols, bases, amines, oxidizing agents. Avoid moisture.

Chemical Identifiers

CAS 623-24-5
Molecular Formula C8H8Br2
Molecular Weight (g/mol) 263.96
MDL Number MFCD00000182
InChI Key RBZMSGOBSOCYHR-UHFFFAOYSA-N
Synonym 1,4-bis bromomethyl benzene, p-xylylene dibromide, alpha,alpha'-dibromo-p-xylene, benzene, 1,4-bis bromomethyl, p-xylylene bromide, p-bromomethyl benzene, 1,4-dibromomethylbenzene, p-bis bromomethyl benzene, 1,4-bis-bromomethyl-benzene, ccris 1775
PubChem CID 69335
IUPAC Name 1,4-bis(bromomethyl)benzene
SMILES C1=CC(=CC=C1CBr)CBr

Specifications

Melting Point 144°C to 148°C
Density 2.012
Boiling Point 245°C
Quantity 10 g
UN Number UN3261
Beilstein 1100019
Solubility Information Soluble in water (react),and toluene.
Formula Weight 263.96
Percent Purity 97%
Chemical Name or Material p-Xylylene dibromide
Hazard Category H314-H318
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 210-781-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only