Learn More
p-Tolylhydrazine hydrochloride, 98%
CAS: 637-60-5 | C7H11ClN2 | 158.629 g/mol
Supplier: Thermo Scientific Chemicals A1422006
Description
p-Tolylhydrazine hydrochloride is used as anti-cancer potential for novel anticancer drugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsp-Tolylhydrazine hydrochloride is used as anti-cancer potential for novel anticancer drugs.
Solubility
Soluble in water, methanol and dimethyl sulfoxide.
Notes
Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.
Specifications
| p-Tolylhydrazine hydrochloride | |
| ∼240°C (decomposition) | |
| MFCD00012940 | |
| UN2811 | |
| p-tolylhydrazine hydrochloride, 4-methylphenylhydrazine hydrochloride, 4-methylphenyl hydrazine hydrochloride, 4-tolylhydrazine hydrochloride, hydrazine, 4-methylphenyl-, monohydrochloride, p-tolylhydrazinium 1+ chloride, hydrazine, p-tolyl-, hydrochloride, 4-methylphenylhydrazine hcl, 1-4-methylphenyl hydrazine hydrochloride, p-tolyl-hydrazine hydrochloride | |
| HMHWNJGOHUYVMD-UHFFFAOYSA-N | |
| (4-methylphenyl)hydrazine;hydrochloride | |
| 12504 | |
| 98% |
| 637-60-5 | |
| C7H11ClN2 | |
| 5 g | |
| 3563996 | |
| Soluble in water,methanol and dimethyl sulfoxide. | |
| CC1=CC=C(C=C1)NN.Cl | |
| 158.629 | |
| 158.63 |
Chemical Identifiers
| 637-60-5 | |
| 158.629 | |
| HMHWNJGOHUYVMD-UHFFFAOYSA-N | |
| 12504 | |
| CC1=CC=C(C=C1)NN.Cl |
| C7H11ClN2 | |
| MFCD00012940 | |
| p-tolylhydrazine hydrochloride, 4-methylphenylhydrazine hydrochloride, 4-methylphenyl hydrazine hydrochloride, 4-tolylhydrazine hydrochloride, hydrazine, 4-methylphenyl-, monohydrochloride, p-tolylhydrazinium 1+ chloride, hydrazine, p-tolyl-, hydrochloride, 4-methylphenylhydrazine hcl, 1-4-methylphenyl hydrazine hydrochloride, p-tolyl-hydrazine hydrochloride | |
| (4-methylphenyl)hydrazine;hydrochloride |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
H302-H315-H319-H335-H351
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 211-295-2
RTECSNumber : MW0195000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only