missing translation for 'onlineSavingsMsg'
Learn More

p-Tolylhydrazine hydrochloride, 98%

Catalog No. p-7021754
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1422006 5 g
AAA1422014 25 g
AAA1422022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1422006 Supplier Thermo Scientific Chemicals Supplier No. A1422006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 637-60-5 | C7H11ClN2 | 158.629 g/mol

p-Tolylhydrazine hydrochloride is used as anti-cancer potential for novel anticancer drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Tolylhydrazine hydrochloride is used as anti-cancer potential for novel anticancer drugs.

Solubility
Soluble in water, methanol and dimethyl sulfoxide.

Notes
Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 637-60-5
Molecular Formula C7H11ClN2
Molecular Weight (g/mol) 158.629
MDL Number MFCD00012940
InChI Key HMHWNJGOHUYVMD-UHFFFAOYSA-N
Synonym p-tolylhydrazine hydrochloride, 4-methylphenylhydrazine hydrochloride, 4-methylphenyl hydrazine hydrochloride, 4-tolylhydrazine hydrochloride, hydrazine, 4-methylphenyl-, monohydrochloride, p-tolylhydrazinium 1+ chloride, hydrazine, p-tolyl-, hydrochloride, 4-methylphenylhydrazine hcl, 1-4-methylphenyl hydrazine hydrochloride, p-tolyl-hydrazine hydrochloride
PubChem CID 12504
IUPAC Name (4-methylphenyl)hydrazine;hydrochloride
SMILES CC1=CC=C(C=C1)NN.Cl

Specifications

Melting Point ∼240°C (decomposition)
Quantity 5 g
UN Number UN2811
Beilstein 3563996
Solubility Information Soluble in water,methanol and dimethyl sulfoxide.
Formula Weight 158.63
Percent Purity 98%
Chemical Name or Material p-Tolylhydrazine hydrochloride
Hazard Category H302-H315-H319-H335-H351
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 211-295-2
RTECSNumber MW0195000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only