missing translation for 'onlineSavingsMsg'
Learn More

p-Tolyldiethanolamine 98.0+%, TCI America™

Catalog No. T079725G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T079725G 25 g
T0797100G 100 g
T0797500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T079725G Supplier TCI America Supplier No. T079725G

Chemical Identifiers

CAS 3077-12-1
Molecular Formula C11H17NO2
Molecular Weight (g/mol) 195.26
MDL Number MFCD00020580
InChI Key JUVSRZCUMWZBFK-UHFFFAOYSA-N
PubChem CID 76497
IUPAC Name 2-[N-(2-hydroxyethyl)-4-methylanilino]ethanol
SMILES CC1=CC=C(C=C1)N(CCO)CCO

Specifications

Melting Point 52°C
Color White-Yellow
Boiling Point 163°C
Quantity 25 g
Synonym N,N-Bis(2-hydroxyethyl)-p-toluidine, 2,2′C-(4-Methylphenylimino)diethanol, 2,2′C-(p-Tolylimino)diethanol
Formula Weight 195.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material p-Tolyldiethanolamine
EINECSNumber (3)-0233
TSCA Yes