accessibility menu, dialog, popup

UserName

CAS RN 3077-12-1

IUPAC Name: 2-[(2-hydroxyethyl)(4-methylphenyl)amino]ethan-1-ol
Synonyms: dihydroxyethyl toluidine
Molecular Weight (g/mol): 195.26
Molecular Formula: C11H17NO2
InChi Key: JUVSRZCUMWZBFK-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)N(CCO)CCO
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results