missing translation for 'onlineSavingsMsg'
Learn More

p-Tolyldiethanolamine 96.0+%, TCI America™

Catalog No. T2455500G
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2455500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T2455500G Supplier TCI America Supplier No. T2455500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3077-12-1
Molecular Formula C11H17NO2
Molecular Weight (g/mol) 195.26
MDL Number MFCD00020580
InChI Key JUVSRZCUMWZBFK-UHFFFAOYSA-N
Synonym N,N-Bis(2-hydroxyethyl)-p-toluidine, 2,2′C-(4-Methylphenylimino)diethanol, 2,2′C-(p-Tolylimino)diethanol
PubChem CID 76497
IUPAC Name 2-[N-(2-hydroxyethyl)-4-methylanilino]ethanol
SMILES CC1=CC=C(C=C1)N(CCO)CCO

Specifications

Melting Point 52°C
Boiling Point 163°C
Quantity 500 g
Formula Weight 195.26
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material p-Tolyldiethanolamine
EINECSNumber (3)-0233
TSCA Yes