missing translation for 'onlineSavingsMsg'
Learn More

p-Tolyl Isobutyrate 97.0+%, TCI America™

Catalog No. I03965ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03965ML 5 mL
I039625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I03965ML Supplier TCI America Supplier No. I03965ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 103-93-5
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.23
MDL Number MFCD00026440
InChI Key UPPSFGGDKACIKP-UHFFFAOYSA-N
Synonym Isobutyric Acid p-Tolyl Ester
PubChem CID 7685
IUPAC Name 4-methylphenyl 2-methylpropanoate
SMILES CC(C)C(=O)OC1=CC=C(C)C=C1

Specifications

Color Yellow
Boiling Point 237°C
Quantity 5 mL
Formula Weight 178.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material p-Tolyl Isobutyrate
RTECSNumber NQ5460000
TSCA Yes