accessibility menu, dialog, popup

UserName

H3CCH3OOCH3

CAS RN 103-93-5

IUPAC Name: 4-methylphenyl 2-methylpropanoate
Synonyms: p-tolyl isobutyrate
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: UPPSFGGDKACIKP-UHFFFAOYSA-N
SMILES: CC(C)C(=O)OC1=CC=C(C)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results