missing translation for 'onlineSavingsMsg'
Learn More

p-Tolyl chloroformate, 97%

Catalog No. ac314560050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC314560050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC314560050 Supplier Thermo Scientific Chemicals Supplier No. 314560050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 937-62-2 | C8H7ClO2 | 170.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent

Chemical Identifiers

CAS 937-62-2
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.6
MDL Number MFCD00013255
InChI Key XOFZPIYYMJUNRG-UHFFFAOYSA-N
Synonym p-tolyl chloroformate, 4-methylphenyl chloroformate, p-tolylchloroformate, tolyl chloroformate, 4-tolylchloroformate, p-tolyl-chloroformate, 4-tolyl chloroformate, p-tolyl carbonochloridate, p-methylphenyl chloroformate
PubChem CID 70304
IUPAC Name (4-methylphenyl) carbonochloridate
SMILES CC1=CC=C(C=C1)OC(=O)Cl

Specifications

Color Colorless
Density 1.1800g/mL
Boiling Point 99.5°C (20.0 mmHg)
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5080 to 1.5100
Linear Formula ClCO2C6H4CH3
Quantity 5 g
Specific Gravity 1.18
Formula Weight 170.6
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material p-Tolyl chloroformate, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
EINECSNumber 213-332-8

RUO – Research Use Only