CAS RN 937-62-2
IUPAC Name:
4-methylphenyl carbonochloridate
Synonyms:
4-methylphenyl carbonochloridate
Molecular Weight (g/mol):
170.59
Molecular Formula:
C8H7ClO2
InChi Key:
XOFZPIYYMJUNRG-UHFFFAOYSA-N
SMILES:
CC1=CC=C(OC(Cl)=O)C=C1
1
–
1
of
1
results
1
p-Tolyl chloroformate, 97%
CAS: 937-62-2 Molecular Formula: C8H7ClO2 Molecular Weight (g/mol): 170.6 MDL Number: MFCD00013255 InChI Key: XOFZPIYYMJUNRG-UHFFFAOYSA-N Synonym: p-tolyl chloroformate,4-methylphenyl chloroformate,p-tolylchloroformate,tolyl chloroformate,4-tolylchloroformate,p-tolyl-chloroformate,4-tolyl chloroformate,p-tolyl carbonochloridate,p-methylphenyl chloroformate PubChem CID: 70304 IUPAC Name: (4-methylphenyl) carbonochloridate SMILES: CC1=CC=C(C=C1)OC(=O)Cl