missing translation for 'onlineSavingsMsg'
Learn More

p-Tolyl Acetate 98.0+%, TCI America™

Catalog No. A067025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A067025G 25 g
A0670500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A067025G Supplier TCI America Supplier No. A067025G

Chemical Identifiers

CAS 140-39-6
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00008703
InChI Key CDJJKTLOZJAGIZ-UHFFFAOYSA-N
Synonym Acetic Acid p-Cresyl Ester, Acetic Acid p-Tolyl Ester, p-Cresyl Acetate
PubChem CID 8797
ChEBI CHEBI:9627
IUPAC Name 4-methylphenyl acetate
SMILES CC(=O)OC1=CC=C(C)C=C1

Specifications

Color Colorless
Boiling Point 214°C
Quantity 25 g
Formula Weight 150.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material p-Tolyl Acetate
EINECSNumber (3)-0632&(3)-0644
RTECSNumber AJ7570000
TSCA Yes