missing translation for 'onlineSavingsMsg'
Learn More

p-Toluic acid, 98%

CAS: 99-94-5 | C8H8O2 | 136.15 g/mol

Supplier:  Thermo Scientific Chemicals A115060I

 View more versions of this product

Catalog No. AAA115060I


Only null left
Explore available promotions

Description

Description

It is an intermediate in some industrial processes for the oxidation of p-xylene to terephthalic acid which is used in the manufacture of polyethylene terephthalate.  It is also used in medicine, agricultural chemicals, API, dye intermediate. p-Toluic Acid is used as an intermediate for polymer stabilizers, light sensitive compounds, animal feed supplements and other organic chemicals (pharmaceuticals, pigments and dyestuff). They are also used as intermediates for the manufacture of a variety of target compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate in some industrial processes for the oxidation of p-xylene to terephthalic acid which is used in the manufacture of polyethylene terephthalate.  It is also used in medicine, agricultural chemicals, API, dye intermediate. p-Toluic Acid is used as an intermediate for polymer stabilizers, light sensitive compounds, animal feed supplements and other organic chemicals (pharmaceuticals, pigments and dyestuff). They are also used as intermediates for the manufacture of a variety of target compounds.

Solubility
It is insoluble in water, but soluble in acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to Strong oxidizing agents, Strong bases. Stable under recommended storage conditions.
Specifications

Specifications

p-Toluic acid
178°C to 182°C
274°C
C8H8O2
5000 g
14,9535
It is insoluble in water,but soluble in acetone.
CC1=CC=C(C=C1)C(O)=O
136.15
CHEBI:36635
98%
99-94-5
1.06
181°C (357°F)
MFCD00002565
507600
p-toluic acid, 4-toluic acid, crithminic acid, p-methylbenzoic acid, p-toluylic acid, benzoic acid, 4-methyl, p-carboxytoluene, para-toluic acid, 4-methyl-benzoic acid, toluenecarboxylic acid
LPNBBFKOUUSUDB-UHFFFAOYSA-N
4-methylbenzoic acid
7470
136.15

Chemical Identifiers

99-94-5
136.15
LPNBBFKOUUSUDB-UHFFFAOYSA-N
7470
4-methylbenzoic acid
C8H8O2
MFCD00002565
p-toluic acid, 4-toluic acid, crithminic acid, p-methylbenzoic acid, p-toluylic acid, benzoic acid, 4-methyl, p-carboxytoluene, para-toluic acid, 4-methyl-benzoic acid, toluenecarboxylic acid
CHEBI:36635
CC1=CC=C(C=C1)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 202-803-3

RTECSNumber : XU1575000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only