missing translation for 'onlineSavingsMsg'
Learn More

p-Toluic acid, 98%

Catalog No. p-201312
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC139061000 100 g Plastic bottle
AC139062500 250 g Plastic bottle
AC139060010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC139061000 Supplier Thermo Scientific Chemicals Supplier No. 139061000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 99-94-5 | C8H8O2 | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99-94-5
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00002565
InChI Key LPNBBFKOUUSUDB-UHFFFAOYSA-N
Synonym p-toluic acid, 4-toluic acid, crithminic acid, p-methylbenzoic acid, p-toluylic acid, benzoic acid, 4-methyl, p-carboxytoluene, para-toluic acid, 4-methyl-benzoic acid, toluenecarboxylic acid
PubChem CID 7470
ChEBI CHEBI:36635
IUPAC Name 4-methylbenzoic acid
SMILES CC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 179.0°C to 182.0°C
Color Cream-Yellow to White
Boiling Point 274.0°C to 275.0°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula CH3C6H4CO2H
Quantity 100 g
Beilstein 09,483
Merck Index 14,9535
Solubility Information Solubility in water: 11.6g/L (98°C). Other solubilities: soluble in alcohol and ether
Formula Weight 136.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material p-Toluic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-803-3
RTECSNumber XU1575000
TSCA TSCA

RUO – Research Use Only