missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Toluenesulfonylacetonitrile 99.0+%, TCI America™
Supplier: TCI America T084625G
Specifications
| p-Toluenesulfonylacetonitrile | |
| 149°C | |
| C9H9NO2S | |
| 25 g | |
| p-toluenesulfonylacetonitrile, 2-tosylacetonitrile, tosylacetonitrile, p-tolylsulfonylacetonitrile, 4-methylphenyl sulfonyl acetonitrile, 4-toluenesulfonylacetonitrile, 4-methylphenyl sulfonylacetonitrile, 4-toluenesulphonylacetonitrile, p-cyanomethylsulfonyl toluene, tosylmethyl isocyanide | |
| CC1=CC=C(C=C1)S(=O)(=O)CC#N | |
| 195.24 | |
| 195.24 | |
| Crystalline Powder |
| 5697-44-9 | |
| White | |
| MFCD00039487 | |
| 3439 | |
| BBNNLJMGPASZPD-UHFFFAOYSA-N | |
| 2-(4-methylbenzenesulfonyl)acetonitrile | |
| 79776 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 5697-44-9 | |
| 195.24 | |
| BBNNLJMGPASZPD-UHFFFAOYSA-N | |
| 79776 | |
| CC1=CC=C(C=C1)S(=O)(=O)CC#N |
| C9H9NO2S | |
| MFCD00039487 | |
| p-toluenesulfonylacetonitrile, 2-tosylacetonitrile, tosylacetonitrile, p-tolylsulfonylacetonitrile, 4-methylphenyl sulfonyl acetonitrile, 4-toluenesulfonylacetonitrile, 4-methylphenyl sulfonylacetonitrile, 4-toluenesulphonylacetonitrile, p-cyanomethylsulfonyl toluene, tosylmethyl isocyanide | |
| 2-(4-methylbenzenesulfonyl)acetonitrile |
Safety and Handling
TSCA : No