accessibility menu, dialog, popup

UserName

CAS RN 5697-44-9

IUPAC Name: 2-(4-methylbenzenesulfonyl)acetonitrile
Synonyms: 2-(4-methylbenzenesulfonyl)acetonitrile
Molecular Weight (g/mol): 195.24
Molecular Formula: C9H9NO2S
InChi Key: BBNNLJMGPASZPD-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)CC#N
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)