missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonylacetonitrile, 98+%

Catalog No. p-7020786
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1285706 5 g
AAA1285714 25 g
AAA1285722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1285706 Supplier Thermo Scientific Chemicals Supplier No. A1285706
Only null left

CAS: 5697-44-9 | C9H9NO2S | 195.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5697-44-9
Molecular Formula C9H9NO2S
Molecular Weight (g/mol) 195.24
MDL Number MFCD00039487
InChI Key BBNNLJMGPASZPD-UHFFFAOYSA-N
Synonym p-toluenesulfonylacetonitrile, 2-tosylacetonitrile, tosylacetonitrile, p-tolylsulfonylacetonitrile, 4-methylphenyl sulfonyl acetonitrile, 4-toluenesulfonylacetonitrile, 4-methylphenyl sulfonylacetonitrile, 4-toluenesulphonylacetonitrile, p-cyanomethylsulfonyl toluene, tosylmethyl isocyanide
PubChem CID 79776
IUPAC Name 2-(4-methylphenyl)sulfonylacetonitrile
SMILES CC1=CC=C(C=C1)S(=O)(=O)CC#N

Specifications

Melting Point 147°C to 152°C
Quantity 5 g
UN Number UN3439
Beilstein 1842131
Formula Weight 195.24
Percent Purity ≥98%
Chemical Name or Material p-Toluenesulfonylacetonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 227-173-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only