missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonyl semicarbazide, 95%

Catalog No. p-7054068
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6194914 25 g
AAH6194922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6194914 Supplier Thermo Scientific Chemicals Supplier No. H6194914
Only null left

CAS: 10396-10-8 | C8H11N3O3S | 229.254 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10396-10-8
Molecular Formula C8H11N3O3S
Molecular Weight (g/mol) 229.254
MDL Number MFCD00072243
InChI Key VRFNYSYURHAPFL-UHFFFAOYSA-N
Synonym p-toluenesulfonylsemicarbazide, 1-tosylsemicarbazide, p-toluenesulfonyl semicarbazide, benzenesulfonic acid, 4-methyl-, 2-aminocarbonyl hydrazide, 1-p-methylphenylsulfonyl semicarbazide, 4-methylbenzenesulfonamidourea, p-toluenesulfohyl semicarbazide, 4-methylbenzenesulfonamido urea, 4-toluenesulfonyl semicarbazide, ra ptss
PubChem CID 82602
IUPAC Name [(4-methylphenyl)sulfonylamino]urea
SMILES CC1=CC=C(C=C1)S(=O)(=O)NNC(=O)N

Specifications

Color White
Quantity 25 g
Formula Weight 229.26
Percent Purity 95%
Physical Form Powder
Chemical Name or Material p-Toluenesulfonyl semicarbazide
Hazard Category Warning
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 233-857-6
TSCA Yes
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only