accessibility menu, dialog, popup

UserName

CAS RN 10396-10-8

IUPAC Name: (4-methylbenzenesulfonamido)urea
Synonyms: 4-methylbenzenesulfonamidourea
Molecular Weight (g/mol): 229.25
Molecular Formula: C8H11N3O3S
InChi Key: VRFNYSYURHAPFL-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)NNC(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results