missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonanilide 98.0+%, TCI America™

Supplier:  TCI America T0282500G

 View more versions of this product

Catalog No. T0282500G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

p-Toluenesulfonanilide
102°C
C13H13NO2S
500 g
VLVCWODDMDGANW-UHFFFAOYSA-N
4-methyl-N-phenylbenzenesulfonamide
2456
≥98.0% (HPLC,T)
68-34-8
White
MFCD00025989
p-toluenesulfonanilide, n-tosylaniline, benzenesulfonamide, 4-methyl-n-phenyl, n-phenyl-p-toluenesulfonamide, 4-methylphenyl sulfonyl phenylamine, p-toluene sulfonanilide, n-phenyl p-toluenesulfonamide, 4-methyl-n-phenyl-benzenesulfonamide, n-4-methylphenyl sulfonyl aniline, 4-methyl-n-phenylbenzene-1-sulfonamide
CC1=CC=C(C=C1)S(=O)(=O)NC2=CC=CC=C2
247.312
247.31
Crystalline Powder

Chemical Identifiers

68-34-8
247.312
VLVCWODDMDGANW-UHFFFAOYSA-N
2456
CC1=CC=C(C=C1)S(=O)(=O)NC2=CC=CC=C2
C13H13NO2S
MFCD00025989
p-toluenesulfonanilide, n-tosylaniline, benzenesulfonamide, 4-methyl-n-phenyl, n-phenyl-p-toluenesulfonamide, 4-methylphenyl sulfonyl phenylamine, p-toluene sulfonanilide, n-phenyl p-toluenesulfonamide, 4-methyl-n-phenyl-benzenesulfonamide, n-4-methylphenyl sulfonyl aniline, 4-methyl-n-phenylbenzene-1-sulfonamide
4-methyl-N-phenylbenzenesulfonamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-3134

RTECSNumber : XT6100000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions