missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonanilide 98.0+%, TCI America™

Catalog No. p-7136187
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T028225G 25 g
T0282500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T028225G Supplier TCI America Supplier No. T028225G
Only null left

Chemical Identifiers

CAS 68-34-8
Molecular Formula C13H13NO2S
Molecular Weight (g/mol) 247.312
MDL Number MFCD00025989
InChI Key VLVCWODDMDGANW-UHFFFAOYSA-N
Synonym p-toluenesulfonanilide, n-tosylaniline, benzenesulfonamide, 4-methyl-n-phenyl, n-phenyl-p-toluenesulfonamide, 4-methylphenyl sulfonyl phenylamine, p-toluene sulfonanilide, n-phenyl p-toluenesulfonamide, 4-methyl-n-phenyl-benzenesulfonamide, n-4-methylphenyl sulfonyl aniline, 4-methyl-n-phenylbenzene-1-sulfonamide
PubChem CID 2456
IUPAC Name 4-methyl-N-phenylbenzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)NC2=CC=CC=C2

Specifications

Melting Point 102°C
Color White
Quantity 25 g
Formula Weight 247.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material p-Toluenesulfonanilide
EINECSNumber (3)-3134
RTECSNumber XT6100000
TSCA Yes