missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Toluenesulfonanilide 98.0+%, TCI America™
Supplier: TCI America T028225G
Specifications
| p-Toluenesulfonanilide | |
| 102°C | |
| C13H13NO2S | |
| 25 g | |
| VLVCWODDMDGANW-UHFFFAOYSA-N | |
| 4-methyl-N-phenylbenzenesulfonamide | |
| 2456 | |
| ≥98.0% (HPLC,T) |
| 68-34-8 | |
| White | |
| MFCD00025989 | |
| p-toluenesulfonanilide, n-tosylaniline, benzenesulfonamide, 4-methyl-n-phenyl, n-phenyl-p-toluenesulfonamide, 4-methylphenyl sulfonyl phenylamine, p-toluene sulfonanilide, n-phenyl p-toluenesulfonamide, 4-methyl-n-phenyl-benzenesulfonamide, n-4-methylphenyl sulfonyl aniline, 4-methyl-n-phenylbenzene-1-sulfonamide | |
| CC1=CC=C(C=C1)S(=O)(=O)NC2=CC=CC=C2 | |
| 247.312 | |
| 247.31 | |
| Crystalline Powder |
Chemical Identifiers
| 68-34-8 | |
| 247.312 | |
| VLVCWODDMDGANW-UHFFFAOYSA-N | |
| 2456 | |
| CC1=CC=C(C=C1)S(=O)(=O)NC2=CC=CC=C2 |
| C13H13NO2S | |
| MFCD00025989 | |
| p-toluenesulfonanilide, n-tosylaniline, benzenesulfonamide, 4-methyl-n-phenyl, n-phenyl-p-toluenesulfonamide, 4-methylphenyl sulfonyl phenylamine, p-toluene sulfonanilide, n-phenyl p-toluenesulfonamide, 4-methyl-n-phenyl-benzenesulfonamide, n-4-methylphenyl sulfonyl aniline, 4-methyl-n-phenylbenzene-1-sulfonamide | |
| 4-methyl-N-phenylbenzenesulfonamide |
Safety and Handling
EINECSNumber : (3)-3134
RTECSNumber : XT6100000
TSCA : Yes