missing translation for 'onlineSavingsMsg'
Learn More

p-Tolualdehyde 98.0+%, TCI America™

Supplier:  TCI America T0259500ML

 View more versions of this product

Catalog No. T0259500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

p-Tolualdehyde
Colorless
C8H8O
500 mL
FXLOVSHXALFLKQ-UHFFFAOYSA-N
4-methylbenzaldehyde
7725
120.15
Liquid
104-87-0
204°C
MFCD00006954
p-tolualdehyde, 4-tolualdehyde, p-formyltoluene, p-tolylaldehyde, p-toluylaldehyde, p-methylbenzaldehyde, para-tolualdehyde, benzaldehyde, 4-methyl, para-methylbenzaldehyde, para-toluyl aldehyde
CC1=CC=C(C=C1)C=O
120.151
CHEBI:28617
≥98.0% (GC)

Chemical Identifiers

104-87-0
120.151
FXLOVSHXALFLKQ-UHFFFAOYSA-N
7725
4-methylbenzaldehyde
C8H8O
MFCD00006954
p-tolualdehyde, 4-tolualdehyde, p-formyltoluene, p-tolylaldehyde, p-toluylaldehyde, p-methylbenzaldehyde, para-tolualdehyde, benzaldehyde, 4-methyl, para-methylbenzaldehyde, para-toluyl aldehyde
CHEBI:28617
CC1=CC=C(C=C1)C=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2659

RTECSNumber : CU7034500

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions