missing translation for 'onlineSavingsMsg'
Learn More

p-Tolualdehyde 98.0+%, TCI America™

Catalog No. p-7136171
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T025925ML 25 mL
T0259100ML 100 mL
T0259500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T025925ML Supplier TCI America Supplier No. T025925ML

Chemical Identifiers

CAS 104-87-0
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.151
MDL Number MFCD00006954
InChI Key FXLOVSHXALFLKQ-UHFFFAOYSA-N
Synonym p-tolualdehyde, 4-tolualdehyde, p-formyltoluene, p-tolylaldehyde, p-toluylaldehyde, p-methylbenzaldehyde, para-tolualdehyde, benzaldehyde, 4-methyl, para-methylbenzaldehyde, para-toluyl aldehyde
PubChem CID 7725
ChEBI CHEBI:28617
IUPAC Name 4-methylbenzaldehyde
SMILES CC1=CC=C(C=C1)C=O

Specifications

Color Colorless
Boiling Point 204°C
Quantity 25 mL
Formula Weight 120.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material p-Tolualdehyde
EINECSNumber (3)-2659
RTECSNumber CU7034500
TSCA Yes