missing translation for 'onlineSavingsMsg'
Learn More

p-Terphenyl (purified by sublimation) 99.5+%, TCI America™

Supplier:  TCI America T326325G

 View more versions of this product

Catalog No. T326325G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

p-Terphenyl (purified by sublimation)
214°C
389°C
MFCD00003061
p-terphenyl, 1,1':4',1-terphenyl, p-diphenylbenzene, p-triphenyl, santowax p, 4-phenylbiphenyl, 4-phenyldiphenyl, para-terphenyl, biphenyl, 4-phenyl, pyrogallol tannin
C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=CC=C3
230.31
CHEBI:52242
≥99.5% (GC)
92-94-4
White
C18H14
25 g
XJKSTNDFUHDPQJ-UHFFFAOYSA-N
1,4-diphenylbenzene
7115
230.31
Crystalline Powder

Chemical Identifiers

92-94-4
230.31
XJKSTNDFUHDPQJ-UHFFFAOYSA-N
7115
1,4-diphenylbenzene
C18H14
MFCD00003061
p-terphenyl, 1,1':4',1-terphenyl, p-diphenylbenzene, p-triphenyl, santowax p, 4-phenylbiphenyl, 4-phenyldiphenyl, para-terphenyl, biphenyl, 4-phenyl, pyrogallol tannin
CHEBI:52242
C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=CC=C3
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0017

RTECSNumber : WZ6475000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions